1-Piperidinepentanol ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 1-(5-Hydroxypentyl)piperidine

P1385-1G P1385-5G
TCP1385-1GEA 72.2 USD
TCP1385-1G TCP1385-5G
1-Piperidinepentanol ≥98.0% (by GC, titration analysis)
1-Piperidinepentanol

CAS Number: 2937-83-9
MDL Number: MFCD03093640
Molecular Formula: C10H21NO
Molecular Weight: 171.28
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 140
Specific Gravity (20/20): 0.94


Other Product Names / Synonyms: 1-(5-Hydroxypentyl)piperidine

Formula: C₁₀H₂₁NO
MW: 171.28 g/mol
Boiling Pt: 140 °C
Density: 0.94 (20 °C)
MDL Number: MFCD03093640
CAS Number: 2937-83-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=64be1de3db49a78447a2ad1e729577606aecd782&vid=64be1de3db49a78447a2ad1e729577606aecd782&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR