Pyridostigmine bromide ≥98.0%

Supplier: TCI America

Synonyms: 3-((Dimethylcarbamoyl)oxy)-1-methylpyridin-1-ium bromide, 3-(Dimethylcarbamoyloxy)-1-methylpyridinium Bromide

P1339-5G P1339-25G
TCP1339-005GEA 37.65 USD
TCP1339-005G TCP1339-025G
Pyridostigmine bromide ≥98.0%
Pyridostigmine bromide

CAS Number: 101-26-8
MDL Number: MFCD00079283
Molecular Formula: C9H13BrN2O2
Molecular Weight: 261.12
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: White
Melting point (°C): 154


Other Product Names / Synonyms: 3-(Dimethylcarbamoyloxy)-1-methylpyridinium Bromide

Formula: C₉H₁₃BrN₂O₂
Melting Pt: 154 °C
MDL Number: MFCD00079283
CAS Number: 101-26-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=012c483ef7463d2e132c0333ba1b166caa0c562a&vid=012c483ef7463d2e132c0333ba1b166caa0c562a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR