Dimethyl-1,4-phenylenediacrylate ≥95.0% (by GC)

Supplier: TCI America

P1338-5G P1338-1G
TCP1338-005GEA 483 USD
TCP1338-005G TCP1338-1G
Dimethyl-1,4-phenylenediacrylate ≥95.0% (by GC)
Dimethyl-1,4-phenylenediacrylate

CAS Number: 7549-44-2
MDL Number: MFCD00185690
Molecular Formula: C14H14O4
Molecular Weight: 246.26
Form: Crystal
Color: Slightly Pale Yellow
Melting point (°C): 167


Other Product Names / Synonyms: 1,4-Phenylenediacrylic Acid Dimethyl Ester

Order Now

Specification Test Results

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 95.0 %
Melting point 166.0 to 170.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR