Phenyl vinyl sulfide ≥98.0% stabilized

Supplier: TCI America

Synonyms: (Phenylthio)ethylene

P1287-5G P1287-25G
TCP1287-005GEA 101 USD
TCP1287-005G TCP1287-25G
Phenyl vinyl sulfide ≥98.0% stabilized
Phenyl vinyl sulfide

(stabilized with TBC)
CAS Number: 1822-73-7
MDL Number: MFCD00009646
Molecular Formula: C8H8S
Molecular Weight: 136.21
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 78
Flash Point (°C): 46
Specific Gravity (20/20): 1.04


Other Product Names / Synonyms: (Phenylthio)ethylene

Formula: C₈H₈S
MW: 136.21 g/mol
Boiling Pt: 78 °C
Density: 1.04 (20 °C)
Flash Pt: 46 °C
MDL Number: MFCD00009646
CAS Number: 1822-73-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4f33f4e1bc0f8ea0b8092b2650a7e6b20d4776a9&vid=4f33f4e1bc0f8ea0b8092b2650a7e6b20d4776a9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR