Diallyl dicarbonate ≥95.0%
Supplier: TCI America
Synonyms:
Diallyl Pyrocarbonate, Pyrocarbonic Acid Diallyl Ester
CAS Number: 115491-93-5
MDL Number: MFCD00077721
Molecular Formula: C8H10O5
Molecular Weight: 186.16
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 93
Specific Gravity (20/20): 1.12
Storage Temperature: 0-10°C
Product Categories: Synthetic Organic Chemistry, Protection & Derivatization Reagents (for Synthesis), Peptide Synthesis, Bioscience, Polyamine Synthesis Reagents
Other Product Names / Synonyms: Diallyl Pyrocarbonate, Pyrocarbonic Acid Diallyl Ester
Formula:
C₈H₁₀O₅ Density: 1.12 (20 °C) Flash Pt: 93 °C |
MDL Number:
MFCD00077721 CAS Number: 115491-93-5 |
|
Specification Test Results
Purity/Analysis Method | >95.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...