Propargyl chloride ∼9.2 mol/L in toluene ≥70%
Supplier: TCI America
Synonyms:
3-Chloropropyne, 3-Chloro-1-propyne
P1273-25G
P1273-250G
TCP1273-025GEA
45
USD
TCP1273-025G
TCP1273-250G
Propargyl chloride ∼9.2 mol/L in toluene ≥70%
Propargyl chloride
CAS Number: 624-65-7
MDL Number: MFCD00000980
Molecular Formula: C3H3Cl
Molecular Weight: 74.51
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 63
Flash Point (°C): -14
Specific Gravity (20/20): 0.98
Other Product Names / Synonyms: 3-Chloro-1-propyne
Formula:
C₃H₃Cl MW: 74.51 g/mol Boiling Pt: 63 °C Density: 0.98 (20 °C) Flash Pt: -14 °C |
MDL Number:
MFCD00000980 CAS Number: 624-65-7 EINECS: 210-856-9 UN: 3286 ADR: 3,I |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...