O-Pentafluorophenyl chlorothioformate ≥95.0% for synthesis

Supplier: TCI America

Synonyms: O-Perfluorophenyl chlorothioformate, Chlorothioformic acid O-Pentafluorophenyl ester, Pentafluorophenyl chlorothionoformate, Pentafluorophenyl Thionochloroformate

P1267-5G P1267-1G
TCP1267-005GEA 202 USD
TCP1267-005G TCP1267-001G
O-Pentafluorophenyl chlorothioformate ≥95.0% for synthesis
O-Pentafluorophenyl chlorothioformate

CAS Number: 135192-53-9
MDL Number: MFCD00075405
Molecular Formula: C7ClF5OS
Molecular Weight: 262.58
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Color: Pale Yellow
Flash Point (°C): 87
Specific Gravity (20/20): 1.64


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: Chlorothioformic Acid O-Pentafluorophenyl Ester, Pentafluorophenyl Thionochloroformate, O-Perfluorophenyl Chlorothioformate

Formula: C₇ClF₅OS
Density: 1.64 (20 °C)
Flash Pt: 87 °C
MDL Number: MFCD00075405
CAS Number: 135192-53-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ead08ecc8cb01b9eaf0e39f73dc4c89c03419658&vid=ead08ecc8cb01b9eaf0e39f73dc4c89c03419658&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR