Tris(trimethylsilyl)phosphate ≥98.0%

Supplier: TCI America

Synonyms: Tris(trimethylsilyl) phosphate, Phosphoric Acid Tris(trimethylsilyl) Ester

P1248-25G P1248-5G
TCP1248-025GEA 189.18 USD
TCP1248-025G TCP1248-5G
Tris(trimethylsilyl)phosphate ≥98.0%
Tris(trimethylsilyl)phosphate

CAS Number: 10497-05-9
MDL Number: MFCD00008267
Molecular Formula: C9H27O4PSi3
Molecular Weight: 314.54
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 236
Flash Point (°C): 24
Specific Gravity (20/20): 0.96


Product Categories: Synthetic Organic Chemistry, Silicon Compounds (for Synthesis)
Other Product Names / Synonyms: Phosphoric Acid Tris(trimethylsilyl) Ester

Formula: C₉H₂₇O₄PSi₃
MW: 314.54 g/mol
Boiling Pt: 236 °C
Density: 0.96 (20 °C)
Flash Pt: 24 °C
MDL Number: MFCD00008267
CAS Number: 10497-05-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1d82f8821e853c1d028499ce28ba6dfedfe9bfe9&vid=1d82f8821e853c1d028499ce28ba6dfedfe9bfe9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR