(S)-(+)-1-(2-Pyrrolidinylmethyl)pyrrolidine ≥98.0%

Supplier: TCI America

P1241-1G P1241-5G
TCP1241-001GEA 133 USD
TCP1241-001G TCP1241-005G
(S)-(+)-1-(2-Pyrrolidinylmethyl)pyrrolidine ≥98.0%
(S)-(+)-1-(2-Pyrrolidinylmethyl)pyrrolidine

CAS Number: 51207-66-0 MDL Number: MFCD00013441 Molecular Formula: C9H18N2 Molecular Weight: 154.26 Purity/Analysis Method: 98.0% (GC,T) Form: Clear Liquid Color: Colorless Boiling point (°C): 84 Flash Point (°C): 87 Specific Gravity (20/20): 0.95 Specific rotation [a]20/D: 10 deg (C=2.4, EtOH)

Formula: C₉H₁₈N₂
Boiling Pt: 84 °C
Density: 0.95 (20 °C)
Flash Pt: 87 °C
MDL Number: MFCD00013441
CAS Number: 51207-66-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5db1846c024d07ce3d50b8b9b1f4a3098bdad386&vid=5db1846c024d07ce3d50b8b9b1f4a3098bdad386&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR