Amoxicillin trihydrate ≥98.0%
Supplier: TCI America
Synonyms:
6-[2-Amino-2-(4-hydroxyphenyl)-acetyl]amino-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid trihydrate, (2S,5R,6R)-6-((R)-2-Amino-2-(4-hydroxyphenyl)acetamido)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid trihydrate, Amoxicillin Ttrihydrate, (2S,5R,6S)-6-((S)-2-amino-2-(4-hydroxyphenyl)acetamido)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid trihydrate
CAS Number: 61336-70-7
MDL Number: MFCD00072029
Molecular Formula: C16H19N3O5S
Molecular Weight: 365.40
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Specific rotation [a]20/D: 302 deg (C=0.1, H2O)
Formula:
C₁₆H₁₉N₃O₅S MW: 365.40 g/mol |
MDL Number:
MFCD00072029 CAS Number: 61336-70-7 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...