2-Pentynyl chloride ≥96.0%
Supplier: TCI America
Synonyms:
1-Chloro-2-pentyne
P1141-25ML
P1141-5ML
TCP1141-025MLEA
1122
USD
TCP1141-025ML
TCP1141-5ML
2-Pentynyl chloride ≥96.0%
2-Pentynyl chloride
CAS Number: 22592-15-0
MDL Number: MFCD00191678
Molecular Formula: C5H7Cl
Molecular Weight: 102.56
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 122
Flash Point (°C): 25
Specific Gravity (20/20): 0.98
Other Product Names / Synonyms: 1-Chloro-2-pentyne
Formula:
C₅H₇Cl MW: 102.56 g/mol Boiling Pt: 122 °C Density: 0.98 (20 °C) Flash Pt: 25 °C |
MDL Number:
MFCD00191678 CAS Number: 22592-15-0 |
Specification Test Results
Purity/Analysis Method | >96.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...