2,4-Dichloro-5-methoxyaniline ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 5-Amino-2,4-dichloroanisole, 4,6-Dichloro-m-anisidine

A2085-25G A2085-5G
TCA2085-25GEA 243 USD
TCA2085-25G TCA2085-5G
2,4-Dichloro-5-methoxyaniline ≥98.0% (by GC, titration analysis)
2,4-Dichloro-5-methoxyaniline

CAS Number: 98446-49-2
MDL Number: MFCD00974410
Molecular Formula: C7H7Cl2NO
Molecular Weight: 192.04
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 51


Other Product Names / Synonyms: 2,4-Dichloro-5-methoxyaniline, 4,6-Dichloro-m-anisidine

Formula: C₇H₇Cl₂NO
Melting Pt: 51 °C
MDL Number: MFCD00974410
CAS Number: 98446-49-2
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7d421851de65ed2f9818069b5165f4c4ba18685a&vid=7d421851de65ed2f9818069b5165f4c4ba18685a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR