Hexadecyl palmitate ≥97.0%
Supplier: TCI America
Synonyms:
Cetyl Palmitate, Palmitic acid palmityl ester, n-Hexadecyl palmitate, Palmitic Acid Hexadecyl Ester, Palmityl palmitate
P1077-25G
P1077-500G
TCP1077-025GEA
63.07
USD
TCP1077-025G
TCP1077-500G
Hexadecyl palmitate ≥97.0%
Hexadecyl palmitate
CAS Number: 540-10-3
MDL Number: MFCD00053739
Molecular Formula: C32H64O2
Molecular Weight: 480.86
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 534
Melting point (°C): 53
Product Categories: Bioscience, Lipids
Other Product Names / Synonyms: Cetyl Palmitate, Palmitic Acid Hexadecyl Ester
Formula:
C₃₂H₆₄O₂ MW: 480.86 g/mol Boiling Pt: 534 °C Melting Pt: 53 °C |
MDL Number:
MFCD00053739 CAS Number: 540-10-3 |
Specification Test Results
Purity/Analysis Method | >97.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...