trans-3-Pentenoic acid ≥95.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: (E)-Pent-3-enoic acid, trans-3-Pentenoic acid

P1072-25ML P1072-5ML
TCP1072-025MLEA 350 USD
TCP1072-025ML TCP1072-5ML
trans-3-Pentenoic acid ≥95.0% (by GC, titration analysis)
trans-3-Pentenoic acid

CAS Number: 1617-32-9 MDL Number: MFCD00063291 Molecular Formula: C5H8O2 Molecular Weight: 100.12 Purity/Analysis Method: 95.0% (GC,T) Form: Clear Liquid Color: Colorless Boiling point (°C): 93 Specific Gravity (20/20): 0.99

Formula: C₅H₈O₂
MW: 100.12 g/mol
Boiling Pt: 93 °C
Density: 0.99 (20 °C)
MDL Number: MFCD00063291
CAS Number: 1617-32-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ca34ba0f82e1269cc1c1d668ab2c6ba96db9f831&vid=ca34ba0f82e1269cc1c1d668ab2c6ba96db9f831&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR