4-Pentyloxyphthalonitrile ≥98.0%
Supplier: TCI America
Synonyms:
1,2-Dicyano-4-pentyloxybenzene, 4-Amyloxyphthalonitrile, 4-(Pentyloxy)phthalonitrile
P1071-1G
P1071-5G
TCP1071-001GEA
63.8
USD
TCP1071-001G
TCP1071-005G
4-Pentyloxyphthalonitrile ≥98.0%
4-Pentyloxyphthalonitrile
CAS Number: 106943-83-3
MDL Number: MFCD00059103
Molecular Formula: C13H14N2O
Molecular Weight: 214.27
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 35
Storage Temperature: 0-10°C
Product Categories: Materials Chemistry, Building Blocks for Phthalocyanines
Other Product Names / Synonyms: 4-Amyloxyphthalonitrile, 1,2-Dicyano-4-pentyloxybenzene
Formula:
C₁₃H₁₄N₂O MW: 214.27 g/mol Melting Pt: 35 °C |
MDL Number:
MFCD00059103 CAS Number: 106943-83-3 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...