Azithromycin dihydrate ≥98.0% (by total nitrogen basis)

Supplier: TCI America

Synonyms: Azithromycin hydrate, (2R,3S,4R,5R,8R,10R,11R,12S,13S,14R)-11-(((2S,3R,4S,6R)-4-(dimethylamino)-3-hydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)-2-ethyl-3,4,10-trihydroxy-13-(((2R,4R,5S,6S)-5-hydroxy-4-methoxy-4,6-dimethyltetrahydro-2H-pyran-2-yl)oxy)-3,5,6,8,10,12,14-heptamethyl-1-oxa-6-azacyclopentadecan-15-one dihydrate

A2076-5G A2076-1G
TCA2076-5GEA 345.17 USD
TCA2076-5G TCA2076-1G
Azithromycin dihydrate ≥98.0% (by total nitrogen basis)
Azithromycin dihydrate

CAS Number: 117772-70-0
MDL Number: MFCD01862248
Molecular Formula: C38H72N2O12
Molecular Weight: 749.00
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Specific rotation [a]20/D: -47 deg (C=2, EtOH)

Formula: C₃₈H₇₂N₂O₁₂·2H₂O
MW: 749.00 g/mol
MDL Number: MFCD01862248
CAS Number: 117772-70-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d869cf528eb71cbc1ecb357ed7646f3156b4cd8b&vid=d869cf528eb71cbc1ecb357ed7646f3156b4cd8b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR