2-Ethylhexyl diphenyl phosphate 90,0
Supplier: TCI America
P1021-25G
P1021-500G
TCP1021-025GEA
37.63
USD
TCP1021-025G
TCP1021-500G
2-Ethylhexyl diphenyl phosphate 90,0
2-Ethylhexyl diphenyl phosphate
CAS Number: 1241-94-7
MDL Number: MFCD00059949
Molecular Formula: C20H27O4P
Molecular Weight: 362.41
Purity/Analysis Method: >90.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 232
Melting point (°C): -30
Flash Point (°C): 224
Specific Gravity (20/20): 1.09
Product Categories: Materials Chemistry, Secondary Battery Research Reagents, Resin Additives
Other Product Names / Synonyms: Octyl Diphenyl Phosphate, Phosphoric Acid Diphenyl 2-Ethylhexyl Ester, Phosphoric Acid 2-Ethylhexyl Diphenyl Ester, Phosphoric Acid Octyl Diphenyl Ester
Formula:
C₂₀H₂₇O₄P MW: 362.3998 g/mol Boiling Pt: 232 °C Melting Pt: –30 °C Density: 1.09 (20 °C) Flash Pt: 224 °C |
MDL Number:
MFCD00059949 CAS Number: 1241-94-7 UN: 3082 ADR: 9 III |
|
Specification Test Results
Purity/Analysis Method | >90.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...