Azathioprine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 6-((1-Methyl-4-nitro-1H-imidazol-5-yl)thio)-9H-purine, 6-(1-Methyl-4-nitroimidazol-5-yl)thiopurine

A2069-25G A2069-5G
TCA2069-25GEA 817.2 USD
TCA2069-25G TCA2069-5G
Azathioprine ≥98.0% (by HPLC, titration analysis)
Azathioprine

CAS Number: 446-86-6
MDL Number: MFCD00069203
Molecular Formula: C9H7N7O2S
Molecular Weight: 277.26
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 244
Storage Temperature: <0°C


Product Categories: Bioscience, Immune System, Inflammation, Allergy Research Reagents, Immunosuppressant Ingredients, Anti-rheumatoid Arthritis Ingredients, Pharmacologically-Active Nucleosides and Nucleobases for Research and Experimental Use, Nucleobases and Their Analogs
Other Product Names / Synonyms: 6-(1-Methyl-4-nitroimidazol-5-yl)thiopurine

Formula: C₉H₇N₇O₂S
MW: 277.26 g/mol
Melting Pt: 244 °C
MDL Number: MFCD00069203
CAS Number: 446-86-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=20d5dba233a4b3d9887df33c2db53cc1f33eb201&vid=20d5dba233a4b3d9887df33c2db53cc1f33eb201&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR