n-Pentacontane ≥98.0%

Supplier: TCI America

P0964-100MG P0964-1G
TCP0964-100MGEA 108 USD
TCP0964-100MG TCP0964-1G
n-Pentacontane ≥98.0%
n-Pentacontane

CAS Number: 6596-40-3
MDL Number: MFCD00042907
Molecular Formula: C50H102
Molecular Weight: 703.37
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 94

Formula: H₃C(CH₂)₄₈CH₃
Melting Pt: 94 °C
Storage Temperature: Ambient
MDL Number: MFCD00042907
CAS Number: 6596-40-3
EINECS: 229-534-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=34f23904e08c5961e0e075f7b619a2c80653e267&vid=d1cda5565c9ec67b78a275b8a6070d93a3c4d570&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR