Nonanoic anhydride ≥97.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Pelargonic Anhydride

P0957-25ML P0957-500ML
TCP0957-025MLEA 66 USD
TCP0957-025ML TCP0957-500ML
Nonanoic anhydride ≥97.0% (by titrimetric analysis)
Nonanoic anhydride

CAS Number: 1680-36-0
MDL Number: MFCD00057768
Molecular Formula: C18H34O3
Molecular Weight: 298.47
Purity/Analysis Method: >97.0% (T)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 207
Specific Gravity (20/20): 0.91


Other Product Names / Synonyms: Pelargonic Anhydride

Formula: C₁₈H₃₄O₃
MW: 298.47 g/mol
Boiling Pt: 207 °C
Density: 0.91 (20 °C)
MDL Number: MFCD00057768
CAS Number: 1680-36-0

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR