Prazosin hydrochloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 1-(4-Amino-6,7-dimethoxy-2-quinazolinyl)-4-(2-furanylcarbonyl)piperazine Hydrochloride, (4-(4-Amino-6,7-dimethoxyquinazolin-2-yl)piperazin-1-yl)(furan-2-yl)methanone hydrochloride, Prazosin HCl

P0938-100MG P0938-1G
TCP0938-100MGEA 69.32 USD
TCP0938-100MG TCP0938-001G
Prazosin hydrochloride ≥98.0% (by titrimetric analysis)
Prazosin hydrochloride

CAS Number: 19237-84-4
MDL Number: MFCD00058177
Molecular Formula: C19H21N5O4
Molecular Weight: 419.87
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 285


Product Categories: Bioscience, Antihypertensive Ingredients
Other Product Names / Synonyms: 1-(4-Amino-6,7-dimethoxy-2-quinazolinyl)-4-(2-furanylcarbonyl)piperazine Hydrochloride

Formula: C₁₉H₂₁N₅O₄
MW: 419.87 g/mol
Melting Pt: 285 °C
MDL Number: MFCD00058177
CAS Number: 19237-84-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e6de62d088f135018c6097674737bbdd417fcac3&vid=e6de62d088f135018c6097674737bbdd417fcac3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR