Vinyl palmitate ≥96.0% stabilized

Supplier: TCI America

P0848-25ML P0848-500ML
TCP0848-025MLEA 86 USD
TCP0848-025ML TCP0848-500ML
Vinyl palmitate ≥96.0% stabilized
Vinyl palmitate

(stabilized with MEHQ)
CAS Number: 693-38-9
MDL Number: MFCD00059288
Molecular Formula: C18H34O2
Molecular Weight: 282.47
Purity/Analysis Method: >96.0% (GC)
Form: Crystal
Flash Point (°C): 176
Freezing point (°C): 22


Other Product Names / Synonyms: Palmitic Acid Vinyl Ester

Formula: C₁₈H₃₄O₂
Flash Pt: 176 °C
MDL Number: MFCD00059288
CAS Number: 693-38-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4bac326d253508edd5a016a7566cd8c55bff7340&vid=4bac326d253508edd5a016a7566cd8c55bff7340&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR