Polyethylene glycol monocetyl ether, M.W. n=∼23

Supplier: TCI America

P0841-500G P0841-25G
TCP0841-500GEA 49.2 USD
TCP0841-500G TCP0841-025G
Polyethylene glycol monocetyl ether, M.W. n=∼23
Polyethylene glycol monocetyl ether

CAS Number: 9004-95-9
MDL Number: MFCD00081419
Form: Wax
Color: White
Flash Point (°C): 150


Other Product Names / Synonyms: Polyethylene Glycol Monohexadecyl Ether, Polyoxyethylene Monocetyl Ether

Formula: HO(CH2CH2O)nC16H33
Flash Pt: 150 °C
MDL Number: MFCD00080892
CAS Number: 9004-95-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=499655a66583312658808d169e2c4d3fa989429e&vid=53221ee376fe3cd03f727d7e0ed5c9c7ef6c8597&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR