Isopropyl propyl sulfide ≥95.0%
Supplier: TCI America
P0826-5ML
P0826-25ML
TCP0826-005MLEA
170
USD
TCP0826-005ML
TCP0826-025ML
Isopropyl propyl sulfide ≥95.0%
Isopropyl propyl sulfide
CAS Number: 5008-73-1
MDL Number: MFCD00027125
Molecular Formula: C6H14S
Molecular Weight: 118.24
Purity/Analysis Method:
Formula:
C₆H₁₄S Boiling Pt: 132 °C Density: 0.83 (20 °C) |
MDL Number:
MFCD00027125 CAS Number: 5008-73-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...