5-Amino-2-methylpyridine ≥98.0%

Supplier: TCI America

Synonyms: 5-Amino-2-picoline, 6-Methylpyridin-3-amine, 3­Amino­6­methylpyridine, 6-Methyl-3-pyridinamine, 3­Amino­6­picoline

A2053-5G A2053-1G
TCA2053-5GEA 63 USD
TCA2053-5G TCA2053-1G
5-Amino-2-methylpyridine ≥98.0%
5-Amino-2-methylpyridine

CAS Number: 3430-14-6
MDL Number: MFCD00833389
Molecular Formula: C6H8N2
Molecular Weight: 108.14
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 95


Other Product Names / Synonyms: 5-Amino-2-picoline

Formula: C₆H₈N₂
MW: 108.14
Melting Pt: 95 °C
MDL Number: MFCD00833389
CAS Number: 3430-14-6
UN: 2811
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=56a803ac091d1874ed6dfd33fcf38c70fd8b0a5b&vid=56a803ac091d1874ed6dfd33fcf38c70fd8b0a5b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR