Propionaldehyde diethyl acetal ≥98.0%
Supplier: TCI America
Synonyms:
1,1-Diethoxypropane, Propionaldehyde Diethyl Acetal
P0781-25ML
P0781-500ML
TCP0781-025MLEA
24
USD
TCP0781-025ML
TCP0781-500ML
Propionaldehyde diethyl acetal ≥98.0%
Propionaldehyde diethyl acetal
CAS Number: 4744-08-5
MDL Number: MFCD00009245
Molecular Formula: C7H16O2
Molecular Weight: 132.20
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 124
Flash Point (°C): 19
Specific Gravity (20/20): 0.83
Other Product Names / Synonyms: 1,1-Diethoxypropane
Formula:
C₇H₁₆O₂ Boiling Pt: 124 °C Density: 0.83 (20 °C) Flash Pt: 19 °C |
MDL Number:
MFCD00009245 CAS Number: 4744-08-5 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...