Lead(II) phthalocyanine ≥95.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Phthalocyanine Lead(II)

P0766-1G P0766-25G
TCP0766-001GEA 51 USD
TCP0766-001G TCP0766-025G
Lead(II) phthalocyanine ≥95.0% (by titrimetric analysis)
Lead(II) phthalocyanine

CAS Number: 15187-16-3
MDL Number: MFCD00053951
Molecular Formula: C32H16N8Pb
Molecular Weight: 719.74
Purity/Analysis Method: >95.0% (T)
Form: Crystal


Product Categories: Materials Chemistry, Molecular Conductors, Solar Cell Research Reagents, Porphyrin & Phthalocyanine Dyes (Functional Organic Dyes), Building Blocks for Phthalocyanines
Other Product Names / Synonyms: Phthalocyanine Lead(II)

Formula: C₃₂H₁₆N₈Pb
MW: 719.74 g/mol
MDL Number: MFCD00053951
CAS Number: 15187-16-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d9cdac882f376523b44a00981c77c0814036910f&vid=d9cdac882f376523b44a00981c77c0814036910f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR