Polyethylene glycol monooleyl ether, M.W. n=∼7
Supplier: TCI America
P0713-25G
P0713-500G
TCP0713-025GEA
19
USD
TCP0713-025G
TCP0713-500G
Polyethylene glycol monooleyl ether, M.W. n=∼7
Polyethylene glycol monooleyl ether
CAS Number: 9004-98-2
MDL Number: MFCD00074369
Form: Clear Liquid
Color: Very Pale Yellow
Specific Gravity (20/20): 0.97
Product Categories: Materials Chemistry, Nonionic Surfactants
Formula:
HO(CH2CH2O)nC18H35 Density: 0.97 (20 °C) |
MDL Number:
MFCD00074369 CAS Number: 9004-98-2 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...