Protamine sulfate Salmon Sperm

Supplier: TCI America

Synonyms: Salmine sulphate

P0675-5G P0675-25G P0675-1G
TCP0675-5GEA 154.38 USD
TCP0675-5G TCP0675-25G TCP0675-1G
Protamine sulfate Salmon Sperm
Protamine sulfate

CAS Number: 53597-25-4
MDL Number: MFCD00132091
Form: Crystal
Color: White
Specific rotation [a]20/D: -70 deg (C=2, H2O)


Other Product Names / Synonyms: Salmine Sulfate

MDL Number: MFCD00132091
CAS Number: 9009-65-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR