Phenylglyoxal monohydrate ≥98.0%

Supplier: TCI America

Synonyms: 2,2-Dihydroxy-1-phenylethan-1-one, Benzoylformaldehyde, Phenylglyoxal hydrate, 2,2-Dihydroxyacetophenone

P0652-5G P0652-25G
TCP0652-005GEA 348 USD
TCP0652-005G TCP0652-025G
Phenylglyoxal monohydrate ≥98.0%
Phenylglyoxal monohydrate

CAS Number: 1075-06-5
MDL Number: MFCD00149499
Molecular Formula: C8H6O2
Molecular Weight: 134.13
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 79


Other Product Names / Synonyms: Benzoylformaldehyde

Formula: C₈H₆O₂·H₂O
Melting Pt: 79 °C
MDL Number: MFCD00149499
CAS Number: 1075-06-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=13e8ac64618630a0e57da7141cdebf00283620e7&vid=13e8ac64618630a0e57da7141cdebf00283620e7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR