Pentamethoxy Red

Supplier: TCI America

Synonyms: Bis(2,4-dimethoxyphenyl)(2-methoxyphenyl)methanol, 2,2',2'',4,4'-Pentamethoxytriphenylmethanol

P0631-1G P0631-25G
TCP0631-001GEA 106.09 USD
TCP0631-001G TCP0631-025G
Pentamethoxy Red
Pentamethoxy Red

CAS Number: 1755-51-7
MDL Number: MFCD00070616
Molecular Formula: C24H26O6
Molecular Weight: 410.47
Form: Crystal
Color: White
Melting point (°C): 146


Other Product Names / Synonyms: Bis(2,4-dimethoxyphenyl)(2-methoxyphenyl)methanol, 2,2',2'',4,4'-Pentamethoxytriphenylmethanol

Formula: C₂₄H₂₆O₆
MW: 410.47 g/mol
Melting Pt: 146 °C
MDL Number: MFCD00070616
CAS Number: 1755-51-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=90b936450eeaad00f8e172757b267208c66cf6e9&vid=90b936450eeaad00f8e172757b267208c66cf6e9&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR