Pontacyl violet 6R
Supplier: TCI America
P0593-1G
P0593-5G
TCP0593-001GEA
130
USD
TCP0593-001G
TCP0593-005G
Pontacyl violet 6R
Pontacyl violet 6R
CAS Number: 5850-63-5
MDL Number: MFCD00059522
Molecular Formula: C20H14N2O8S2
Molecular Weight: 518.42
Form: Crystal
Color: Purple
Formula:
C₂₀H₁₄N₂O₈S₂ |
MDL Number:
MFCD00059522 CAS Number: 5850-63-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...