Piperonyl butoxide ≥90.0%

Supplier: TCI America

Synonyms: 5-[2-(2-Butoxyethoxy)ethoxymethyl]-6-propyl-1,3-benzodioxole, 5-((2-(2-Butoxyethoxy)ethoxy)methyl)-6-propylbenzo[d][1,3]dioxole, NSC 8401

P0458-25G P0458-500G
TCP0458-025GEA 22 USD
TCP0458-025G TCP0458-500G
Piperonyl butoxide ≥90.0%
Piperonyl butoxide

CAS Number: 51-03-6
MDL Number: MFCD00005842
Molecular Formula: C19H30O5
Molecular Weight: 338.44
Purity/Analysis Method: >90.0% (GC)
Form: Clear Liquid
Boiling point (°C): 195
Flash Point (°C): 171
Specific Gravity (20/20): 1.06


Other Product Names / Synonyms: 5-[2-(2-Butoxyethoxy)ethoxymethyl]-6-propyl-1,3-benzodioxole

Formula: C₁₉H₃₀O₅
Boiling Pt: 195 °C
Density: 1.06 (20 °C)
Flash Pt: 171 °C
MDL Number: MFCD00005842
CAS Number: 51-03-6
UN: 3082
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=03570cdb5717efab3f6941e7625e11e99e790666&vid=03570cdb5717efab3f6941e7625e11e99e790666&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >90.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR