Methyl picolinate ≥98.0%

Supplier: TCI America

Synonyms: Methyl pyridine-2-carboxylate

P0423-25G P0423-250G
TCP0423-025GEA 53 USD
TCP0423-025G TCP0423-250G
Methyl picolinate ≥98.0%
Methyl picolinate

CAS Number: 2459-07-6
MDL Number: MFCD00038038
Molecular Formula: C7H7NO2
Molecular Weight: 137.14
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 233
Flash Point (°C): 103
Freezing point (°C): 19
Specific Gravity (20/20): 1.17


Other Product Names / Synonyms: Methyl Picolinate, Picolinic Acid Methyl Ester, Pyridine-2-carboxylic Acid Methyl Ester

Formula: C₇H₇NO₂
MW: 137.14 g/mol
Boiling Pt: 233 °C
Density: 1.17 (20 °C)
Flash Pt: 103 °C
MDL Number: MFCD00038038
CAS Number: 2459-07-6
EINECS: 219-545-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=09499943ea53ccd880818d88a3e9b456f408c68a&vid=09499943ea53ccd880818d88a3e9b456f408c68a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR