Phytin
Supplier: TCI America
P0410-500G
P0410-25G
TCP0410-500GEA
148.6
USD
TCP0410-500G
TCP0410-025G
Phytin
Phytin
CAS Number: 3615-82-5
MDL Number: MFCD00082315
Form: Crystal
Color: White
Other Product Names / Synonyms: Calcium Phytate
MDL Number:
MFCD00082315 CAS Number: 3615-82-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...