L-Phenylephrine ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
(R)-(-)-1-(3-Hydroxyphenyl)-2-(methylamino)ethanol, (R)-(-)-3-[1-Hydroxy-2-(methylamino)ethyl]phenol, (R)-(-)-3-Hydroxy-alpha-(methylaminomethyl)benzyl Alcohol, (R)-(-)-Phenylephrine
CAS Number: 59-42-7
MDL Number: MFCD00044749
Molecular Formula: C9H13NO2
Molecular Weight: 167.21
Purity/Analysis Method:
Product Categories: Bioscience, Phenylethylamines
Other Product Names / Synonyms: (R)-(-)-Phenylephrine, (R)-(-)-3-[1-Hydroxy-2-(methylamino)ethyl]phenol, (R)-(-)-1-(3-Hydroxyphenyl)-2-(methylamino)ethanol, (R)-(-)-3-Hydroxy-alpha-(methylaminomethyl)benzyl Alcohol
Formula:
C₉H₁₃NO₂ MW: 167.21 g/mol Melting Pt: 174 °C |
MDL Number:
MFCD00044749 CAS Number: 59-42-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...