Poly(1-vinylpyrrolidone-co-vinyl acetate) (copolymer 7:3) 50% in ethanol

Supplier: TCI America

P0382-25G P0382-500G
TCP0382-025GEA 20 USD
TCP0382-025G TCP0382-500G
Poly(1-vinylpyrrolidone-co-vinyl acetate) (copolymer 7:3) 50% in ethanol
Poly(1-vinylpyrrolidone-co-vinyl acetate)

CAS Number: 25086-89-9
MDL Number: MFCD00134018
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 13


Product Categories: Materials Chemistry, Polymers

Flash Pt: 13 °C
MDL Number: MFCD00134018
CAS Number: 25086-89-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR