Phenylthiohydantoin-δ-threonine ≥90.0%

Supplier: TCI America

Synonyms: PTH-DELTA-threonine, 5-Ethylidene-3-phenyl-2-thiohydantoin

P0370-1G P0370-100MG
TCP0370-001GEA 193 USD
TCP0370-001G TCP0370-100MG
Phenylthiohydantoin-δ-threonine ≥90.0%
Phenylthiohydantoin-delta-threonine

CAS Number: 5800-50-0
MDL Number: MFCD00070617
Molecular Formula: C11H10N2OS
Molecular Weight: 218.27
Purity/Analysis Method: >90.0% (HPLC)
Form: Crystal
Melting point (°C): 236


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: 5-Ethylidene-3-phenyl-2-thiohydantoin, PTH-DELTA-threonine

Formula: C₁₁H₁₀N₂OS
Melting Pt: 236 °C
MDL Number: MFCD00070617
CAS Number: 5800-50-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e66a5483dac06c5c90e5b037da1f6a1e3acd7897&vid=e66a5483dac06c5c90e5b037da1f6a1e3acd7897&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >90.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR