2-Aminoethyl dihydrogen phosphate ≥98.0% for synthesis
Supplier: TCI America
Synonyms:
O-Phosphorylethanolamine
P0279-25G
P0279-100G
TCP0279-025GEA
105.4
USD
TCP0279-025G
TCP0279-100G
2-Aminoethyl dihydrogen phosphate ≥98.0% for synthesis
2-Aminoethyl dihydrogen phosphate
CAS Number: 1071-23-4
MDL Number: MFCD00008178
Molecular Formula: C2H8NO4P
Molecular Weight: 141.06
Purity/Analysis Method: >98.0% (N,T)
Form: Crystal
Melting point (°C): 235
Other Product Names / Synonyms: O-Phosphoethanolamine, 2-Aminoethyl Dihydrogen Phosphate
Formula:
C₂H₈NO₄P MW: 141.0630 g/mol Melting Pt: 235 °C |
MDL Number:
MFCD00008178 CAS Number: 1071-23-4 EINECS: 213-988-5 UN: 3261 ADR: 8 II |
|
Specification Test Results
Purity/Analysis Method | >98.0% (N,T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...