Tris(1,3-dichloro-2-propyl)phosphate ≥95.0%
Supplier: TCI America
Synonyms:
Phosphoric Acid Tris(1,3-dichloro-2-propyl) Ester
P0269-25G
P0269-500G
TCP0269-025GEA
43.74
USD
TCP0269-025G
TCP0269-500G
Tris(1,3-dichloro-2-propyl)phosphate ≥95.0%
Tris(1,3-dichloro-2-propyl)phosphate
CAS Number: 13674-87-8
MDL Number: MFCD00083121
Molecular Formula: C9H15Cl6O4P
Molecular Weight: 430.89
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 252
Flash Point (°C): 249
Specific Gravity (20/20): 1.52
Product Categories: Materials Chemistry, Secondary Battery Research Reagents, Resin Additives, Resin Additives
Other Product Names / Synonyms: Phosphoric Acid Tris(1,3-dichloro-2-propyl) Ester
Formula:
C₉H₁₅Cl₆O₄P Boiling Pt: 252 °C Density: 1.52 (20 °C) Flash Pt: 249 °C |
MDL Number:
MFCD00083121 CAS Number: 13674-87-8 |
|
Specification Test Results
Purity/Analysis Method | >95.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...