Triallyl phosphate ≥96.0% (by GC)
Supplier: TCI America
Synonyms:
Phosphoric Acid Triallyl Ester
P0264-25ML
P0264-100ML
TCP0264-025MLEA
145
USD
TCP0264-025ML
TCP0264-100ML
Triallyl phosphate ≥96.0% (by GC)
Triallyl phosphate
CAS Number: 1623-19-4
MDL Number: MFCD00014946
Molecular Formula: C9H15O4P
Molecular Weight: 218.19
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 113
Specific Gravity (20/20): 1.08
Product Categories: Materials Chemistry, Secondary Battery Research Reagents, Monomers
Other Product Names / Synonyms: Phosphoric Acid Triallyl Ester
Formula:
C₉H₁₅O₄P MW: 218.19 g/mol Boiling Pt: 113 °C Density: 1.08 (20 °C) |
MDL Number:
MFCD00014946 CAS Number: 1623-19-4 EINECS: 216-606-5 |
Specification Test Results
Appearance | Colourless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...