6-Amino-2,4-dichloro-3-methylphenol hydrochloride ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: 6-Amino-2,4-dichloro-m-cresol Hydrochloride

A1998-25G A1998-5G
TCA1998-25GEA 189.32 USD
TCA1998-25G TCA1998-5G
6-Amino-2,4-dichloro-3-methylphenol hydrochloride ≥98.0% (by HPLC)
6-Amino-2,4-dichloro-3-methylphenol hydrochloride

CAS Number: 39549-31-0
MDL Number: MFCD00035472
Molecular Formula: C7H7Cl2NO
Molecular Weight: 228.50
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal


Other Product Names / Synonyms: 6-Amino-2,4-dichloro-m-cresol Hydrochloride

Formula: C₇H₇Cl₂NO
MW: 228.50 g/mol
MDL Number: MFCD00035472
CAS Number: 39549-31-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=96cac2bdf7af1829d190aac4538a3298867c46d2&vid=96cac2bdf7af1829d190aac4538a3298867c46d2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR