2-Ethylhexyl phosphate (mono and di ester mixture)

Supplier: TCI America

P0261-500G P0261-25G
TCP0261-500GEA 140.9 USD
TCP0261-500G TCP0261-025G
2-Ethylhexyl phosphate (mono and di ester mixture)
2-Ethylhexyl phosphate (mono and di ester mixture)

CAS Number: 12645-31-7
MDL Number: MFCD00135884
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 142
Specific Gravity (20/20): 1.01


Other Product Names / Synonyms: Phosphoric Acid 2-Ethylhexyl Ester, Octyl Phosphate, Phosphoric Acid Octyl Ester

Density: 1.01 (20 °C)
Flash Pt: 142 °C
MDL Number: MFCD00135884
CAS Number: 12645-31-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR