Phosphoenolpyruvic acid tris(cyclohexylammonium)salt, hydrate ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
PEP-3CHA
P0256-1G
P0256-5G
TCP0256-1GEA
151
USD
TCP0256-1G
TCP0256-5G
Phosphoenolpyruvic acid tris(cyclohexylammonium)salt, hydrate ≥98.0% (by titrimetric analysis)
Phosphoenolpyruvic acid tris(cyclohexylammonium)salt
CAS Number: 35556-70-8
MDL Number: MFCD00004258
Molecular Formula: C3H5O6P
Molecular Weight: 465.57
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Storage Temperature: 0-10°C
Formula:
C₃H₅O₆P·3C₆H₁₄N MW: 465.57 g/mol |
MDL Number:
MFCD00004258 CAS Number: 35556-70-8 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...