1-Phenyl-3-thiosemicarbazide ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

P0234-25G P0234-1G
TCP0234-025GEA 182 USD
TCP0234-025G TCP0234-001G
1-Phenyl-3-thiosemicarbazide ≥98.0% (by HPLC, titration analysis)
1-Phenyl-3-thiosemicarbazide

CAS Number: 645-48-7
MDL Number: MFCD00042739
Molecular Formula: C7H9N3S
Molecular Weight: 167.23
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 196

Formula: C₇H₉N₃S
Melting Pt: 196 °C
MDL Number: MFCD00042739
CAS Number: 645-48-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4f3415fda3c38989168eeba495aba498fbe64e84&vid=4f3415fda3c38989168eeba495aba498fbe64e84&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR