3-Phenylsalicylic acid ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
2-Hydroxy-[1,1'-biphenyl]-3-carboxylic acid, 2-Hydroxybiphenyl-3-carboxylic Acid
P0226-25G
P0226-5G
TCP0226-025GEA
256
USD
TCP0226-025G
TCP0226-5G
3-Phenylsalicylic acid ≥98.0% (by titrimetric analysis)
3-Phenylsalicylic acid
CAS Number: 304-06-3
MDL Number: MFCD00020257
Molecular Formula: C13H10O3
Molecular Weight: 214.22
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 191
Other Product Names / Synonyms: 2-Hydroxybiphenyl-3-carboxylic Acid
Formula:
C₁₃H₁₀O₃ MW: 214.22 g/mol Melting Pt: 191 °C |
MDL Number:
MFCD00020257 CAS Number: 304-06-3 |
Specification Test Results
Purity/Analysis Method | >98.0% (T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...