2'-Aminobenzophenone-2-carboxylic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 2'-Aminobenzophenone-2-carboxylic Acid, 2-Anthraniloylbenzoic acid, 2-(2-Aminobenzoyl)benzoic acid

A1994-5G A1994-1G
TCA1994-5GEA 122 USD
TCA1994-5G TCA1994-1G
2'-Aminobenzophenone-2-carboxylic acid ≥98.0% (by HPLC, titration analysis)
2'-Aminobenzophenone-2-carboxylic acid

CAS Number: 1147-43-9
MDL Number: MFCD00007715
Molecular Formula: C14H11NO3
Molecular Weight: 241.25
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 199


Other Product Names / Synonyms: 2-(2-Aminobenzoyl)benzoic Acid, 2-Anthraniloylbenzoic Acid

Formula: C₁₄H₁₁NO₃
MW: 241.2420 g/mol
Melting Pt: 199 °C
MDL Number: MFCD00007715
CAS Number: 1147-43-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=00fbdfe682737e653ea6544a994aedb4fb767ae1&vid=00fbdfe682737e653ea6544a994aedb4fb767ae1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR