2-Phenylphenol sodium salt tetrahydrate ≥80.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 2-Hydroxybiphenyl Sodium Salt, Sodium 2-Phenylphenolate, Sodium 2-Hydroxybiphenyl, 2-Biphenylol Sodium Salt, Sodium o-Phenylphenate, Sodium 2-Biphenylolate

P0202-500G P0202-25G
TCP0202-500GEA 121 USD
TCP0202-500G TCP0202-025G
2-Phenylphenol sodium salt tetrahydrate ≥80.0% (by titrimetric analysis)
2-Phenylphenol sodium salt tetrahydrate

CAS Number: 132-27-4
MDL Number: MFCD00002209
Molecular Formula: C12H10O
Molecular Weight: 192.19
Purity/Analysis Method: >80.0% (T)
Form: Crystal
Melting point (°C): 78


Other Product Names / Synonyms: 2-Hydroxybiphenyl Sodium Salt, Sodium 2-Hydroxybiphenyl, Sodium 2-Phenylphenolate, 2-Biphenylol Sodium Salt, Sodium 2-Biphenylolate

Formula: C₁₂H₁₀O
MW: 192.19 g/mol
Melting Pt: 78 °C
MDL Number: MFCD00002209
CAS Number: 6152-33-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=387fb18027cb6ac0b581d36f3ee985d01896d871&vid=387fb18027cb6ac0b581d36f3ee985d01896d871&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >80.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR