o-Phenylenediamine dihydrochloride ≥98.0% for synthesis

Supplier: TCI America
Danger

Synonyms: Benzene-1,2-diamine dihydrochloride, 1,2-Phenylenediammonium dichloride, 1,2-Diaminobenzene dihydrochloride, 1,2-Phenylenediamine dihydrochloride

P0171-500G P0171-25G
TCP0171-500GEA 572 USD
TCP0171-500G TCP0171-025G
o-Phenylenediamine dihydrochloride ≥98.0% for synthesis
o-Phenylenediamine dihydrochloride

CAS Number: 615-28-1
MDL Number: MFCD00012966
Molecular Formula: C6H8N2
Molecular Weight: 181.06
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 205


Other Product Names / Synonyms: 1,2-Diaminobenzene Dihydrochloride

Formula: C₆H₁₀Cl₂N₂
MW: 181.06 g/mol
Melting Pt: 205
MDL Number: MFCD00012966
CAS Number: 615-28-1
EINECS: 210-418-7
UN: 2811
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1cc9eb36ec30fd16209c981809c3d7e7f1bdc6bf&vid=1cc9eb36ec30fd16209c981809c3d7e7f1bdc6bf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR