4-Phenyl-1-butene ≥98.0%
Supplier: TCI America
P0161-5ML
P0161-25ML
TCP0161-005MLEA
87.34
USD
TCP0161-005ML
TCP0161-025ML
4-Phenyl-1-butene ≥98.0%
4-Phenyl-1-butene
CAS Number: 768-56-9
MDL Number: MFCD00026111
Molecular Formula: C10H12
Molecular Weight: 132.21
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 181
Flash Point (°C): 53
Specific Gravity (20/20): 0.88
Formula:
C₁₀H₁₂ Boiling Pt: 181 °C Density: 0.88 (20 °C) Flash Pt: 53 °C |
MDL Number:
MFCD00026111 CAS Number: 768-56-9 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...